Seeker/Probe, Electron Microscopy Sciences | Avantor
Seeker/Probe, Electron Microscopy Sciences
Double End Seeker/Probe
Catalog # 100493-912
Fournisseur #:  72945-02
Seeker/Probe, Electron Microscopy Sciences
Catalog # 100493-912
Seeker/Probe, Electron Microscopy Sciences
Seeker/Probe, Electron Microscopy Sciences
Double End Seeker/Probe
Price
27,75 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel with a hexagonal handle
Single end probe which comes with one fine point curved tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip.
Spécifications
  • Désignation:
    Double End Seeker/Probe