Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, 20×
  100491-464
Supplier #:  68148
Steinheil Magnifier, Electron Microscopy Sciences
  100491-464
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, 20×
$258.61
Each
The magnifier helps the user eliminate bulky and heavier conventional magnifiers.
A biconvex lens. Pocket size. The sliding frame is designed for the storage of the lens after use. It is good for the preliminary inspection of microscopic specimens. It is lightweight and has excellent chromatic correction with a reasonably wide field of view. Size: 39 x 24 x 22 mm. Average weight: 20 g.
  • Grossissement:
    20×
  • Désignation:
    Steinheil Magnifier