Large Glass Slides, Electron Microscopy Sciences
Large Glass Slides, Electron Microscopy Sciences
75x38 mm glass slide, 1 mm thick
Catalog # 100491-372
Supplier #:  71860-01
Large Glass Slides, Electron Microscopy Sciences
Catalog # 100491-372
Large Glass Slides, Electron Microscopy Sciences
Large Glass Slides, Electron Microscopy Sciences
75x38 mm glass slide, 1 mm thick
Price
$212.64
Log in pour la disponibilité
Each Gross of 144
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These slides are precleaned, individually selected and are suitable for large specimen sections, such as, brain tissue.
In addition to our comprehensive collection of glass slides manufactured by ESCO we now carry large glass slides which are available in 75×38 mm and 75×51 mm.
Spécifications
  • Désignation:
    75x38 mm glass slide, 1 mm thick