Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Product Details & Documents
Good for handling infectious materials, inoculating culture media and routine sampling. Available in either Borosilicate or Flint glass, with an outside body diameter of 7 mm.
Specifications
- Désignation:Borosilicate
- Longueur:4"L
- Cat. No.:100490-958