CoverWell™ Imaging Chambers, Electron Microscopy Sciences
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
- Profondeur:0.9 mm
- Désignation:1 well, 300 µl
- Taille:20 mm dia