Bench Top Covers, Electron Microscopy Sciences | Avantor
Bench Top Covers, Electron Microscopy Sciences
Bench Top Covers Moderate Absorbency, 17" x 24"
Catalog # 100490-584
Fournisseur #:  64456-05
Bench Top Covers, Electron Microscopy Sciences
Catalog # 100490-584
Bench Top Covers, Electron Microscopy Sciences
Bench Top Covers, Electron Microscopy Sciences
Bench Top Covers Moderate Absorbency, 17" x 24"
Price
82,93 $
Log in pour la disponibilité
Each Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A wide selection of bench top covers from economy layered to premium ultra absorbent.
Spécifications
  • Désignation:
    Bench Top Covers Moderate Absorbency
  • Taille:
    17" x 24"