Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences | Avantor
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
125 ml (4.2 oz.)
Catalog # 100490-552
Fournisseur #:  64298-04
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Catalog # 100490-552
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
125 ml (4.2 oz.)
Price
$142.07
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette. The pipettes are graduated for nominal indication of amount of liquid being drawn. Bellows-shaped bulb makes for easy use and provides excellent drop information.
Spécifications
  • Capacité:
    125 ml (4.2 oz.)