Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Product Details & Documents
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
Spécifications
- Ø int.:32 mm
- Capacité:15 Dram (55.5 ml)
- Hauteur:64 mm