Dropping Bottles, Glass, Electron Microscopy Sciences | Avantor
Dropping Bottles, Glass, Electron Microscopy Sciences
Amber Dropping Bottle, 100 ml
Catalog # 100490-432
Fournisseur #:  64221-10
Dropping Bottles, Glass, Electron Microscopy Sciences
Catalog # 100490-432
Dropping Bottles, Glass, Electron Microscopy Sciences
Amber Dropping Bottle, 100 ml
Price
$316.63
Log in pour la disponibilité
Each Pack of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper.
A partial turn of the ground glass stopper will seal the bottle.
Spécifications
  • Désignation:
    Amber Dropping Bottle, 100 ml