Emery Paper, Electron Microscopy Sciences
Emery paper, 4/0 grit, 800 mesh
  100490-134
Emery Paper, Electron Microscopy Sciences
  100490-134
 :  
Emery Paper, Electron Microscopy Sciences
Emery Paper, Electron Microscopy Sciences
Emery paper, 4/0 grit, 800 mesh
This emery paper comes in sheets and in varying grit sizes.
Grit 4/0 is the finest and 3 is the coarsest.
Size: 22×34 cm (9×13¾")
  • Désignation:
    Emery paper, 4/0 grit, 800 mesh