Perfect Loop, Electron Microscopy Sciences | Avantor
Perfect Loop, Electron Microscopy Sciences
Perfect Loop Only
Catalog # 100489-830
Fournisseur #:  70946
Perfect Loop, Electron Microscopy Sciences
Catalog # 100489-830
Perfect Loop, Electron Microscopy Sciences
Perfect Loop, Electron Microscopy Sciences
Perfect Loop Only
Price
$1,355.35
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.
Spécifications
  • Désignation:
    Perfect Loop Only