Dispensing Bottles, Electron Microscopy Sciences
60 ml (2 oz.)
  100489-770
Dispensing Bottles, Electron Microscopy Sciences
  100489-770
Dispensing Bottles, Electron Microscopy Sciences
Dispensing Bottles, Electron Microscopy Sciences
60 ml (2 oz.)
$66.75
Each Pack of 10
Low density polyethylene bottles.
  • Tip has a 0.76 mm (0.003") opening
By squeezing the bottle a steady stream of liquid is produced.
  • Capacité:
    60 ml (2 oz.)