The Original LKB Grid Storage Box, Electron Microscopy Sciences | Avantor
The Original LKB Grid Storage Box, Electron Microscopy Sciences
LKB 100 Grid Box
Catalog # 100489-238
Fournisseur #:  71147-12
The Original LKB Grid Storage Box, Electron Microscopy Sciences
Catalog # 100489-238
The Original LKB Grid Storage Box, Electron Microscopy Sciences
The Original LKB Grid Storage Box, Electron Microscopy Sciences
LKB 100 Grid Box
Price
$1,060.43
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The box  consists of 100 diamond shaped holes for storing up to 100 EM grids, either 3.05 or 2.3 mm in diameter.
  • The box measures L x W x H 75 x 55 x 7 mm (3 x 2 1/8 x 1/4")
  • It comes complete with 10 index cards
For years, The LKB Grid Storage Box is the one that everyone is looking for. Now it is available again from EMS. The box is made from ABS (a copolymer of Acrylonitrile, Butadien and Styrene) which will not tolerate temperatures above 70 °C, while the lid is made of  Polymethacrylate (Flexiglas, Perspex), which should not be exposed to temperatures above 45 °C. Neither the box or the lid will resist organic solvents.
Spécifications
  • Désignation:
    LKB 100 Grid Box