PTFE Printed Slides, Electron Microscopy Sciences
PTFE Printed Slides, Electron Microscopy Sciences
Hydrophobic printed slides, 30 well, 2 mm
  100488-912
Supplier #:  63434-02
PTFE Printed Slides, Electron Microscopy Sciences
  100488-912
PTFE Printed Slides, Electron Microscopy Sciences
PTFE Printed Slides, Electron Microscopy Sciences
Hydrophobic printed slides, 30 well, 2 mm
$332.24
Each Pack of 72
These slides are extremely hydrophobic, acetone proof, chemical resistant and autoclavable. They are perfect for controlling cross-contamination in staining, immunolabeling and immunofluorescence tests.
  • Super hydrophobic mask
  • Clean, wettable wells
  • A bioadhesive surface treatment to enhance cell and tissue attachment
The well surface is 100% wettable, which enhances cell attachment and increases its spreading capabilities within the well. These slides minimize the use of expensive purified antigens and antibodies. The slides measure 3 x 1" (76 x 26 x 1 mm).
Manufactured to order offering maximum shelf life. The shelf life of 18 months to two years. However, it is recommended that all glass slides (Printed or Un-printed) be used in a timely manner to avoid glass "weathering", (slides becoming cloudy or hazy even with proper storage. All printed slides are autoclavable and have a frosted end. They are intended for one time use only.
  • Désignation:
    Hydrophobic printed slides, 30 well
  • L×l:
    2 mm