Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Carbon Rods, Electron Microscopy Sciences | Avantor

Carbon Rods, Electron Microscopy Sciences

304.8 mm (12"), 3.2 mm (1/8"), Carbon (Graphite) Rods Spectro-Grade
Catalog # 100488-370
Fournisseur #:  70200
Carbon Rods, Electron Microscopy Sciences
Catalog # 100488-370
Carbon Rods, Electron Microscopy Sciences
Carbon Rods, Electron Microscopy Sciences

304.8 mm (12"), 3.2 mm (1/8"), Carbon (Graphite) Rods Spectro-Grade

Price
$320.06
Log in pour la disponibilité
Each Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Carbon /graphite films are ideal in EM due to uniform amorphous form and high transparency to electron beams. Carbon/graphite rods are CVP™ (Chemical Vapor Process) purity and spectrographic grade, grade 1, produced to ASTM tolerances. Spectro-Grade rods are ideal for all TEM, SEM and all other critical applications.

  • Spectro-Grade graphite rods have an impurity level of 2 ppm or less with a maximum level element of each of 1 ppm
  • Technical grade carbon has a typical impurity level of 50 ppm or greater
  • For non critical SEM coating applications, technical grade may be used

Graphite is the stable form of carbon. Graphite is one of the softest and diamond is one of the hardest minerals known to man. Carbon is harder than graphite. For TEM and SEM applications, graphite is recommended.

Carbon /graphite films are most usable in TEM and SEM due to its value characteristics: uniformity amorphous and high transparent to electron beams. Because of low molecular weight and their unique structure (graphite have a sheet like structure, where the atoms all lie in a plane), carbon/graphite films have excellent mechanical stability, and even film thickness is about 1 – 2 nm.

Spécifications
  • Ø:
    3.2 mm (1/8")
  • Désignation:
    Carbon (Graphite) Rods Spectro-Grade
  • Longueur:
    304.8 mm (12")

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.