Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark pen, Assorted
  101413-504
Supplier #:  72127-AS
Manomark Pen All Surface Marker, Electron Microscopy Sciences
  101413-504
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark pen, Assorted
$365.15
Each Pack of 12
Writes on all plastic, glass, metal, wool, paper and cloth, including "PTFE", fluoropolymer resins
  • With its precision seal valve the pens life is preserved and as well it allows for a smooth flow of ink even if it’s left uncapped
  • Once dry, the solvent base ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
Markings are insoluble in water or hydrocarbons, resist many solvents, and are easily removed with alcohol.
  • Color:
    Assorted
  • Désignation:
    Manomark pen