Xylene Substitute, Electron Microscopy Sciences | Avantor
Xylene Substitute, Electron Microscopy Sciences
Xylene substitute, Bottle, 5 gal.
Catalog # 100504-384
Fournisseur #:  23410-05
Xylene Substitute, Electron Microscopy Sciences
Catalog # 100504-384
CAS Number:  
Xylene Substitute, Electron Microscopy Sciences
Xylene substitute, Bottle, 5 gal.
CAS Number:  
Price
1 264,94 $
Log in pour la disponibilité
Each (5US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This EMS Xylene Substitute solution is specially designed for use in processing and staining histological/cytological specimens. It is essentially odorless with low toxicity, less irritating to the skin than xylene- and d-limonene-based clearants. It can be substituted for xylene and can be used with almost all tissue processors and automated slide stainers.
  • Minimal tissue shrinkage
  • Dries slowly with no residue
  • Less toxic than xylene
  • Soluble with alcohol, paraffin embedding media, and mounting media
  • Biodegradable
Xylene substitute for histology and cytology is a safer alternative to xylene. Can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. It can be used in all procedures that require xylene.
Spécifications
Spécifications
  • Type de conditionnement:
    Bottle
  • Désignation:
    Xylene substitute
  • Taille:
    5 gal.