Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks (Stainless Steel), 0.1"
  100503-046
Supplier #:  50152-02
Flat Mounting Blocks , Electron Microscopy Sciences
  100503-046
Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks (Stainless Steel), 0.1"
$166.21
Each
Stainless steel flat mounting blocks with the diameter of 25 mm (1") are used for lapping and polishing fixtures.
It is used for the models 150, 151 and 155.
  • Ø:
    0.1"
  • Désignation:
    Flat Mounting Blocks (Stainless Steel)