Automatic Pipette Washing Set, Electron Microscopy Sciences | Avantor
Automatic Pipette Washing Set, Electron Microscopy Sciences
Pipette Jar
Catalog # 100502-542
Fournisseur #:  64377-15
Automatic Pipette Washing Set, Electron Microscopy Sciences
Catalog # 100502-542
Automatic Pipette Washing Set, Electron Microscopy Sciences
Automatic Pipette Washing Set, Electron Microscopy Sciences
Pipette Jar
Price
$467.29
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Solve the problem of washing pipettes quickly, safely, and with little effort. To use allow dirty pipettes to soak in the pipette basket in the pipette jar and when ready the basket is transferred to the washer where a constant flow of tap water rinses the pipettes clean.
Spécifications
  • Désignation:
    Pipette Jar