Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Disque en carbure de silicium, 8", 120, PSA,
  100500-926
 :  50255-03
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
  100500-926
 :  
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Disque en carbure de silicium, 8", 120, PSA,
Silicone Carbide disc are the finest wet or dry abrasive discs for sample preparation
Wheels are available in 9 different grit sizes, ranging from coarse to fine, on plain back or water resistant pressure sensitive adhesive back.
Pressure sensitive adhesive backed discs eliminates the need to transfer to plates. Silicon Carbide Abrasive Cut-Off Wheels offered long life, and affordably priced sectioning. All wheels are 9" (228.6 mm) diameter 0.062” (1.6 mm) thick; 1¼” (31mm) arbor.
 : Available in 8" diameter discs as a standard and 12" discs available upon request.
  • Désignation:
    Disque en carbure de silicium, 8"
  • Backing:
    PSA
  • Grit:
    120