Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences | Avantor
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Catalog # 100500-900
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Catalog # 100500-900
Supplier:  
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
CRYOVIAL PP NS 2.0ML BK CS1000 Catalog #66008-748
Le produit 66008-748 est similaire à 100500-900 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures.
  • Graduated in 0.5 ml increments
The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal.