Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences | Avantor
Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Lab Tek glass slide with 2 chambers, 6
Catalog # 100500-602
Fournisseur #:  70394
Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Catalog # 100500-602
Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Lab Tek glass slide with 2 chambers, 6
Price
$1,544.86
Log in pour la disponibilité
Box of 6 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Permanox® is a special molding resin, which meets the growing requirements of most anchorage dependent cells; also excellent chemical resistance allows for cell fixation and the staining of organic solvents for EM and other techniques. Low oxygen permeation makes Permanox® ideal for anoxic cell growth.
  • Developed for microscopy of in situ cell culture
  • Cells grow on a standard microscope slide
  • Media chamber can be removed when culturingis complete
  • Useful for viral and mycoplasma testing, chromosome studies toxicity tests and immunocytology
  • Permanox® plastic and glass do not interfere with fluorescent labels
The chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox® plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded using our Slide Duplicating Mold (Cat. 70170). Lab-Tek® Chamber Slide™ System all come packed in a sterile sealed tray, 8 units per tray, 2 trays per pack and 12 trays per case.
Spécifications
  • Désignation:
    Lab Tek glass slide with 2 chambers
  • No. of Trays:
    6