British Standard Graticules - NG10, Electron Microscopy Sciences
British Standard Graticules - NG10, Electron Microscopy Sciences
NG10 Graticule, 16 mm
Catalog # 100499-242
Supplier #:  68026-16
British Standard Graticules - NG10, Electron Microscopy Sciences
Catalog # 100499-242
British Standard Graticules - NG10, Electron Microscopy Sciences
British Standard Graticules - NG10, Electron Microscopy Sciences
NG10 Graticule, 16 mm
Price
774,46 $
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Originally designed by the National Coal Board for use in coal mining.
In this graticule the circle areas double progressively, hence the diameters alter by Root 2, so that the size classes can form a continuation of the standard series of sieves for particle sizing. Each particle is assigned to a size class defined by two adjacent circles, which represent the size limits of that class. Thus the distribution of size is obtained in terms of the diameter of circles having the same projected area as the particles. This method will cover particles in the range 150 micron to 0.38 micron. The size distributions with respect to their number and weight are determined separately. Final results are calculated as cumulative percents.
Specifications
  • Désignation:
    NG10 Graticule
  • Taille:
    16 mm