Top-Loading Dispensing Bins, Electron Microscopy Sciences
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Small Top-Loading Dispensing Bins
Catalog # 100498-774
Supplier #:  64388-05
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Catalog # 100498-774
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Small Top-Loading Dispensing Bins
Price
$298.73
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bins are made from ¹/₄" thick PVC. They are chemical resistant and easy to clean.
  • Removable top for easy loading
  • Hinged clear acrylic door
  • Different sizes for your needs
Ideal for pipette tips, small vials, cassette lids, cassette base molds, cotton swabs, aluminum stubs, embedding capsules etc.
Specifications
  • Désignation:
    Small Top-Loading Dispensing Bins