Automatic Pipette Washing Set, Electron Microscopy Sciences
Pipette Basket
  100498-752
 :  64377-25
Automatic Pipette Washing Set, Electron Microscopy Sciences
  100498-752
Automatic Pipette Washing Set, Electron Microscopy Sciences
Automatic Pipette Washing Set, Electron Microscopy Sciences
Pipette Basket
$375.21
Each
Solve the problem of washing pipettes quickly, safely, and with little effort. To use allow dirty pipettes to soak in the pipette basket in the pipette jar and when ready the basket is transferred to the washer where a constant flow of tap water rinses the pipettes clean.
  • Désignation:
    Pipette Basket