Tray Set with Plastic Boxes, Electron Microscopy Sciences | Avantor
Tray Set with Plastic Boxes, Electron Microscopy Sciences
Tray Set With Plastic Boxes, 56 Box Set
Catalog # 100498-678
Fournisseur #:  64350-06
Tray Set with Plastic Boxes, Electron Microscopy Sciences
Catalog # 100498-678
Tray Set with Plastic Boxes, Electron Microscopy Sciences
Tray Set with Plastic Boxes, Electron Microscopy Sciences
Tray Set With Plastic Boxes, 56 Box Set
Price
212,32 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A compact, solid fiberboard tray box makes instituting a plastic box/sample storage system more convenient and efficient.
  • Available with or without accompanying plastic boxes
To form a tray set, use plastic boxes for specimen storage, and then organize boxes within the fiberboard tray Box. These sets eliminates misplacement or inadvertent sample loss and allows for rapid retrieval of specimens.
Spécifications
  • Désignation:
    Tray Set With Plastic Boxes
  • Taille:
    56 Box Set