Diamond Points, Electron Microscopy Sciences
Diamond Points, Tapered, 7.3 mm, Wheel
Catalog # 100497-988
 :  62155-73
Diamond Points, Electron Microscopy Sciences
Catalog # 100497-988
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Tapered, 7.3 mm, Wheel
Price
65,77 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Spécifications
  • Ø:
    7.3 mm
  • Désignation:
    Diamond Points, Tapered
  • Shape:
    Wheel