Abrasive Wheels, Electron Microscopy Sciences
Product Details & Documents
Finishing and Polishing Wheels and Discs.
Pumice Wheels have 72 Assortment Wheels. For cleaning, polishing and finishing. Made from natural rubber with graded pumice grain distributed to assure uniform results. Kit includes 12 discs each of six styles and 3 mandrels with 3.18 mm (¹/₈") shanks.
Sanding Discs are yellow abrasive with press on/flip-off center hole feature. Aluminum oxide grit in three grades, mounted on a thin backing, provides outstanding results in sanding and smoothing operations. Individual Discs: Sold 100 discs in a tube. Assortment Disc: Contains 25 fine, 25 coarse, and 50 medium, 2.2 mm (⁷/₈") sanding discs, plus one mandrel, in a plastic compartment box.
Press-On/Flip-Off Mandrel is made from high quality steel with 2.35 mm (³/₃₂") shank.
The resilient rubber binder cushions the aluminum oxide abrasive and bridges the gap between grinding and polishing. 57 piece kit contains wheels cylinders, points, mandrels and chuck.
Specifications
- Désignation:Press On/Flip Off Mandrell