Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, 71 mm id
  100496-218
Stabilizing Rings, Electron Microscopy Sciences
  100496-218
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, 71 mm id
$148.46
Each
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
  • Désignation:
    Stabilizing Ring, 71 mm id