Sample Storage Sets; Square Bottle, Electron Microscopy Sciences | Avantor
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Square Bottle Series, 232.00 ml, Oui
Catalog # 100496-190
Fournisseur #:  64284
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Catalog # 100496-190
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Square Bottle Series, 232.00 ml, Oui
Price
$285.47
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A large size storage set. It comes with square clear glass bottles (1 ounce capacity) and black screw caps with plastic linings.
Alpha-numerical indexing system allows quick identification and location of stored items.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Spécifications
  • Avec bouchon:
    Oui
  • Capacité:
    232.00 ml
  • Désignation:
    Square Bottle Series