High Opacity Paint Markers, Electron Microscopy Sciences
Large Marker, Red
  100493-636
 :  72167-R2
High Opacity Paint Markers, Electron Microscopy Sciences
  100493-636
High Opacity Paint Markers, Electron Microscopy Sciences
High Opacity Paint Markers, Electron Microscopy Sciences
Large Marker, Red
$16.62
Each
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface
  • Weatherproof
  • Resistant to alcohol and acetone
  • Contain xylene
Markers are available in black, red and white with bullet tips. Except model 130, it writes like a pen with white ink for marking on dark surfaces, including carbon tabs, which is use in SEM specimen mounting purpose.
  • Désignation:
    Large Marker, Red