Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
20-Slide Staining Dish and Lid only
Catalog # 100493-304
Supplier #:  71420-DL
Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
Catalog # 100493-304
Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
20-Slide Staining Dish and Lid only
Price
$185.80
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These glass staining dishes are made of a heavy glass with snug fit lids.
  • Snug fit lids are included
  • Dishes have gasket seal
The stainless steel rack has a swing handle for easy access and has grooves to take 76 mm microscope slides, up to 40 mm wide horizontally.
Inside Dimensions: 33/4 x 3 x 21/4" (H)
Outside Dimension: 41/4 x 35/8 x 3" (H)
Delivery information: Complete set includes one staining dish with lid and one swing handle rack.
Specifications
  • Désignation:
    20-Slide Staining Dish and Lid only