BEEM™ Dial-A-Grid and Block Storage Modules, Electron Microscopy Sciences | Avantor
BEEM™ Dial-A-Grid and Block Storage Modules, Electron Microscopy Sciences
Dial-A-Grid & Block Storage
Catalog # 100493-138
Fournisseur #:  71149-01
BEEM™ Dial-A-Grid and Block Storage Modules, Electron Microscopy Sciences
Catalog # 100493-138
BEEM™ Dial-A-Grid and Block Storage Modules, Electron Microscopy Sciences
BEEM™ Dial-A-Grid and Block Storage Modules, Electron Microscopy Sciences
Dial-A-Grid & Block Storage
Price
$19.97
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
The same as Dial-A-Grid Module but with two additional cavities for block storage.
Spécifications
  • Désignation:
    Dial-A-Grid & Block Storage