Clip-on Magnifiers, Electron Microscopy Sciences
Clip On Magnifier, 1.5×, 2, 508 mm (20")
Catalog # 100492-784
 :  68260-02
Clip-on Magnifiers, Electron Microscopy Sciences
Catalog # 100492-784
Supplier:  
Clip-on Magnifiers, Electron Microscopy Sciences
Clip-on Magnifiers, Electron Microscopy Sciences
Clip On Magnifier, 1.5×, 2, 508 mm (20")
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clips on to any eyeglass frame leaving hands free.
Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
Spécifications
  • Grossissement:
    1.5×
  • Désignation:
    Clip On Magnifier
  • Diopters:
    2
  • Working Distance:
    508 mm (20")