Scotch Conductive Tapes, Electron Microscopy Sciences
Product Details & Documents
Copper and aluminum conductive tapes with continuous long term usage up to 155 °C
- Functional days-weeks usage up to 175 °C
- Copper double sided available
We offer two types of foil: Aluminum foil tape (Scotch 1170) and copper foil tape (Scotch 1181) with conductive adhesive. 3” core, 1/4" (6.35 mm) width×54 ft (16.5 m) long.
Spécifications
- Largeur:¼"
- Désignation:Conductive Aluminum Tape