Scintillation Vial Rack, Electron Microscopy Sciences
Product Details & Documents
Welded polypropylene rack holds 20 ml scintillation vials.
- Autoclavable
- Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
Specifications
- Désignation:Scintillation vial rack with handle
- No. of Places:24
- W×D×H:11¾×5×3¾"