Forensic Collection Kit, Electron Microscopy Sciences
Forensic Collection Kit, Electron Microscopy Sciences
Kit with Plastic Vial, 23.5 mm dx52.5 mm h
Catalog # 100491-318
Supplier #:  76480-03
Forensic Collection Kit, Electron Microscopy Sciences
Catalog # 100491-318
Supplier:  
Forensic Collection Kit, Electron Microscopy Sciences
Forensic Collection Kit, Electron Microscopy Sciences
Kit with Plastic Vial, 23.5 mm dx52.5 mm h
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Manufactured from a special – certified aluminum alloy, containing only trace amounts of Barium (
These kits are suitable for field work and gunshot residue in the lab or out.
Manufactured from a special – certified aluminum alloy, containing only trace amounts of Barium (~0.00010%), Copper (~0.0139%), Zinc (~0.0094%), Nickel (~0.00309%), and Antimony (~0.00080%). These sample mounts are suitable for typical forensic sample studies.
The kit consists of 10 pin mounts (12.74 mm diameter) 10 carbon adhesive tabs, 10 storage vials, and one plastic tweezer. Two styles of glass vials are available as well as one in plastic. 0.00010%), Copper (
0.0139%), Zinc (
0.0094%), Nickel (
0.00309%), and Antimony (
0.00080%). These sample mounts are suitable for typical forensic sample studies.
The kit consists of 10 pin mounts (12.74 mm diameter) 10 carbon adhesive tabs, 10 storage vials, and one plastic tweezer. Two styles of glass vials are available as well as one in plastic.
Specifications
  • Désignation:
    Kit with Plastic Vial, 23.5 mm dx52.5 mm h