Forensic Collection Kit, Electron Microscopy Sciences
Product Details & Documents
Manufactured from a special – certified aluminum alloy, containing only trace amounts of Barium (
These kits are suitable for field work and gunshot residue in the lab or out.
Manufactured from a special – certified aluminum alloy, containing only trace amounts of Barium (~0.00010%), Copper (~0.0139%), Zinc (~0.0094%), Nickel (~0.00309%), and Antimony (~0.00080%). These sample mounts are suitable for typical forensic sample studies.
The kit consists of 10 pin mounts (12.74 mm diameter) 10 carbon adhesive tabs, 10 storage vials, and one plastic tweezer. Two styles of glass vials are available as well as one in plastic. 0.00010%), Copper (
0.0139%), Zinc (
0.0094%), Nickel (
0.00309%), and Antimony (
0.00080%). These sample mounts are suitable for typical forensic sample studies.
The kit consists of 10 pin mounts (12.74 mm diameter) 10 carbon adhesive tabs, 10 storage vials, and one plastic tweezer. Two styles of glass vials are available as well as one in plastic.
Spécifications
- Désignation:Kit with Glass Shell Vial, 23.7 mmx37 mm h