Carolina™ Blue Box, Electron Microscopy Sciences
Product Details & Documents
Molded from blue high-impact styrene, the Carolina Slide Box is manufactured to exacting specifications.
Compared to conventional wooden boxes, the Carolina Blue Box is smaller in size, lighter in weight, and held to closer dimensional tolerances. The 75 x 25 mm (3 x 1") slides fit the slots and will not slip out once in place. A shoulder on each box excludes dust and identities the top of the box. The boxes are available in two sizes: 3 1/4 x 4 1/4 x 1 3/16" (for 12 slides).
Spécifications
- Capacité:25 Slide
- Désignation:Carolina Blue Box