BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences | Avantor
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial-A-Grid Box
Catalog # 100490-974
Fournisseur #:  71148-05
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Catalog # 100490-974
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial-A-Grid Box
Price
$813.41
Log in pour la disponibilité
Each Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A two tone color coded plastic box with insert which has 24 letter-labeled crossed slots, where the grids can be stored. A rotating protection plate covers the slots and allows for exposure of one grid at a time. Measures L x W x H: 57 x 45 x 12.5 mm (2 1/3 x 1 3/4 x 1/2").
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Spécifications
  • Désignation:
    Dial-A-Grid Box