Disposable Glass Tubes, Electron Microscopy Sciences
Yellow Label, 100 mm, 13 mm
  100490-622
 :  64636-Y
Disposable Glass Tubes, Electron Microscopy Sciences
  100490-622
Disposable Glass Tubes, Electron Microscopy Sciences
Disposable Glass Tubes, Electron Microscopy Sciences
Yellow Label, 100 mm, 13 mm
$629.63
Each Pack of 1,000
Tubes are made from borosilicate glass, round bottom and plain top.
The tubes measuring 16 mm in outer diameter and 125 mm in length are Flint glass only.
Labeling area in five pastel colors. Handy fused-in color patches, accept any type of ink or pencil.
  • Ø:
    13 mm
  • Désignation:
    Yellow Label
  • Hauteur:
    100 mm