Transport/Sample Collection Vials and Tubes, Electron Microscopy Sciences
Transport vials/tubes are ideal for sample collection and storage.
- Conical tubes with freestanding skirts
- Tubes are autoclavable
- Write-on area on side
Transport Vials are made from virgin polypropylene. They are available in three sizes, and come either sterile or non sterile. Graduations provide convenient and accurate references.
Transport Tubes are made from virgin polypropylene (PP) or polystyrene (PS). They also work well in Olympus automated testing equipment. Plug caps are required.
: 5 ml and 10 ml vials are supplied with white screw cap and 50 mL vials are supplied with blue cap that has a molded-in sealing ring for a leak-proof fit. Plug caps are available in yellow color and you can choose from hollow top (HT) or hollow bottom (HB).
- Volume:12 ml
- Désignation:Conical Bottom Tube, PP, No Cap