Spec-Cloth, Electron Microscopy Sciences | Avantor
Spec-Cloth, Electron Microscopy Sciences
Spec-Cloth Polishing Cloth, PSA, 21/2" centered hole
Catalog # 100490-008
Fournisseur #:  50320-15
Spec-Cloth, Electron Microscopy Sciences
Catalog # 100490-008
Spec-Cloth, Electron Microscopy Sciences
Spec-Cloth, Electron Microscopy Sciences
Spec-Cloth Polishing Cloth, PSA, 21/2" centered hole
Price
$117.83
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A long napped rayon flock with a plastic barrier.
This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Available with Pressure Sensitive Adhesive (PSA) with and without hole.
Spécifications
  • Désignation:
    Spec-Cloth Polishing Cloth, PSA, 21/2" centered hole