Spec-Cloth, Electron Microscopy Sciences
Product Details & Documents
A long napped rayon flock with a plastic barrier.
This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Available with Pressure Sensitive Adhesive (PSA) with and without hole.
Spécifications
- Désignation:Spec-Cloth Polishing Cloth, PSA no hole