Tooled Neck Vials, Electron Microscopy Sciences | Avantor
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, 2DR, Oui
Catalog # 100489-524
Fournisseur #:  72646-02
Tooled Neck Vials, Electron Microscopy Sciences
Catalog # 100489-524
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, Electron Microscopy Sciences
Tooled Neck Vials, 2DR, Oui
Price
$218.94
Log in pour la disponibilité
Each Pack of 144
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.
Spécifications
  • Avec bouchon:
    Oui
  • Désignation:
    Tooled Neck Vials, 2DR