SEM Conductive Tapes, Electron Microscopy Sciences | Avantor
SEM Conductive Tapes, Electron Microscopy Sciences
Double Sided Carbon Tape
Catalog # 100488-832
Fournisseur #:  77817
SEM Conductive Tapes, Electron Microscopy Sciences
Catalog # 100488-832
SEM Conductive Tapes, Electron Microscopy Sciences
SEM Conductive Tapes, Electron Microscopy Sciences
Double Sided Carbon Tape
Price
$524.60
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
We now offer a new line of conductive tapes, that are electrically conductive and have a clean background. They were developed especially for scanning electron microscopy.
Spécifications
  • Désignation:
    Double Sided Carbon Tape