4-Compartment Micromesh Cassettes, Electron Microscopy Sciences | Avantor
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
White
Catalog # 101413-226
Fournisseur #:  70075-W
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Catalog # 101413-226
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
White
Price
$800.43
Log in pour la disponibilité
Each Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Biopsy processing cassettes with four square compartments each measuring 13 mm.
  • Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
Spécifications
  • Color:
    White