Serum Bottles, Electron Microscopy Sciences
Product Details & Documents
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
Certifications: Meets USP Type I standards.
Spécifications
- Capacité:50 ml
- Color:Clear
- Désignation:Serum Bottles