MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
Product Details & Documents
Scissors are made from 20 gauge stainless steel and have a 3 mm cutting edge.
Specifications
- Désignation:Horizontal Opening
- Curvature:Curved