Large Glass Slides, Electron Microscopy Sciences
Product Details & Documents
These slides are precleaned, individually selected and are suitable for large specimen sections, such as, brain tissue.
In addition to our comprehensive collection of glass slides manufactured by ESCO we now carry large glass slides which are available in 75×38 mm and 75×51 mm.
Spécifications
- Désignation:75x51 mm glass slide, 1.2 mm thick